Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

136980

Sigma-Aldrich

α2-Chloroisodurene

98%

Synonym(s):

2,4,6-Trimethylbenzyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3C6H2CH2Cl
CAS Number:
Molecular Weight:
168.66
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

solid

mp

39-42 °C (lit.)

functional group

chloro

SMILES string

Cc1cc(C)c(CCl)c(C)c1

InChI

1S/C10H13Cl/c1-7-4-8(2)10(6-11)9(3)5-7/h4-5H,6H2,1-3H3

InChI key

UNRGEIXQCZHICP-UHFFFAOYSA-N

General description

α2-Chloroisodurene undergoes coupling reaction with methylmagnesium iodide to yield 1,2-dimesitylethane[1].

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

222.8 °F - closed cup

flash_point_c

106 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Coupling Action of the Grignard Reagent. VII. 1 Di-o-methyl Benzyl Chlorides.
Fuson RC, et al.
Journal of the American Chemical Society, 63(10), 2652-2653 (1941)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service