Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13476

Sigma-Aldrich

(+)-Dimethyl 2,3-O-benzylidene-D-tartrate

≥98.0% (sum of enantiomers, GC)

Synonym(s):

(4S,5S)-2-Phenyl-1,3-dioxolane-4,5-dicarboxylic acid dimethyl ester, Dimethyl (4S,5S)-2-phenyl-1,3-dioxolane-4,5-dicarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14O6
CAS Number:
Molecular Weight:
266.25
Beilstein/REAXYS Number:
4704640
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

≥98.0% (sum of enantiomers, GC)

form

solid

optical activity

[α]20/D +33±1°, c = 7% in benzene

mp

73-75 °C

storage temp.

2-8°C

SMILES string

COC(=O)[C@H]1OC(O[C@@H]1C(=O)OC)c2ccccc2

InChI

1S/C13H14O6/c1-16-11(14)9-10(12(15)17-2)19-13(18-9)8-6-4-3-5-7-8/h3-7,9-10,13H,1-2H3/t9-,10-/m0/s1

InChI key

FMSYNRGOFIGJMM-UWVGGRQHSA-N

Other Notes

Starting material for the synthesis of chiral building blocks[1]; Preparation of 2-O-benzyl-D-threitol and protected D-threose derivative[2][3][4]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K. Fumita et al.
Tetrahedron Letters, 23, 3507-3507 (1982)
G. Adam et al.
Synthesis, 373-373 (1988)
D.R. Williams et al.
Tetrahedron Letters, 28, 869-869 (1987)
D. Seebach et al.
Modern Synthetic Methods, 2, 91-91 (1980)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service