Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13423

Sigma-Aldrich

(S)-(+)-Glycidyl benzyl ether

puriss., ≥99.0% (sum of enantiomers, GC)

Synonym(s):

(S)-(+)-2-(Benzyloxymethyl)oxirane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O2
CAS Number:
Molecular Weight:
164.20
Beilstein/REAXYS Number:
5246494
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

grade

puriss.

Quality Level

assay

≥99.0% (sum of enantiomers, GC)

form

solid

optical activity

[α]20/D +5.1±0.5°, c = 5% in toluene

refractive index

n20/D 1.517

density

1.072 g/mL at 20 °C (lit.)

SMILES string

C(OCc1ccccc1)[C@@H]2CO2

InChI

1S/C10H12O2/c1-2-4-9(5-3-1)6-11-7-10-8-12-10/h1-5,10H,6-8H2/t10-/m1/s1

InChI key

QNYBOILAKBSWFG-SNVBAGLBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(S)-(+)-Glycidyl benzyl ether is aryl glycidyl ether. Enantiomerically pure aryl and benzyl glycidyl ethers have been reported to undergo stereospecific cyclizations, affording 3-chromanols or tetrahydrobenzo[c]oxepin-4-ols.[1] (S)-benzyl glycidyl ether has been produced with 30%ee along with (R)-3-benzyloxypropane-1,2-diol with 40%ee, via its resolution using whole cells of Bacillus alcalophilus.[2]

Application

(S)-(+)-Glycidyl benzyl ether may be employed for the total asymmetric synthesis of (+)-gigantecin.[3]

Other Notes

Important chiral building block reacting with nucleophiles at C-3 of the oxirane ring[4][5][6][7][8][9][10][11]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T.M. Avignon et al.
Tetrahedron, 47, 7279-7279 (1991)
Journal of the Chemical Society. Chemical Communications, 1371-1371 (1989)
S. Takano et al.
Synthesis, 539-539 (1989)
Rocío Marcos et al.
Journal of the American Chemical Society, 130(50), 16838-16839 (2008-12-05)
Enantiomerically pure aryl and benzyl glycidyl ethers undergo stereospecific cyclizations leading to 3-chromanols or to tetrahydrobenzo[c]oxepin-4-ols under mild conditions in the presence of a catalytic amount of FeBr3/3AgOTf. The same processes are also mediated, albeit in a much less efficient
R.E. Ireland et al.
The Journal of Organic Chemistry, 55, 1423-1423 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service