Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13410

Sigma-Aldrich

Benzyldimethyl(2-hydroxyethyl)ammonium chloride

≥97.0% (AT)

Synonym(s):

Benzyl(2-hydroxyethyl)dimethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2N(Cl)(CH3)2CH2CH2OH
CAS Number:
Molecular Weight:
215.72
Beilstein/REAXYS Number:
6896989
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

≥97.0% (AT)

form

solid

mp

75-78 °C

SMILES string

[Cl-].C[N+](C)(CCO)Cc1ccccc1

InChI

1S/C11H18NO.ClH/c1-12(2,8-9-13)10-11-6-4-3-5-7-11;/h3-7,13H,8-10H2,1-2H3;1H/q+1;/p-1

InChI key

NUPDFKWZQURWCC-UHFFFAOYSA-M

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service