Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

132012

Sigma-Aldrich

m-Toluidine

99%

Synonym(s):

3-Methylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4NH2
CAS Number:
Molecular Weight:
107.15
Beilstein/REAXYS Number:
635944
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

assay

99%

refractive index

n20/D 1.567 (lit.)

bp

203-204 °C (lit.)

density

0.999 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(N)c1

InChI

1S/C7H9N/c1-6-3-2-4-7(8)5-6/h2-5H,8H2,1H3

InChI key

JJYPMNFTHPTTDI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

m-Toluidine (MT) is an aromatic amine. It has been reported as a fragile molecular glass-forming liquid.[1] Mechanical αα-relaxation process of MT has been described.[2]

Application

m-Toluidine may be used in the preparation of following:
  • Pyrrole(PY)/m-toluidine (MT) copolymers, via oxidative polymerization.[3]
  • La/Sm-doped strontium-ferrite/poly-m-toluidine) (PMT) composites.4
  • Pyrrole and m-toluidine copolymer (P(PY/MT))/montmorillonite (MMT) composites.[4]
  • La-doped Barium-ferrite/Poly-m-toluidine composites.6

Legal Information

DuPont product

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The α-relaxation process in simple glass forming liquid m-toluidine. II. The temperature dependence of the mechanical response.
Cutroni M and Mandanici A.
J. Chem. Phys. , 114(16), 7124-7129 (2001)
Synthesis and characterization of pyrrole and m-toluidine copolymers.
Li XG, et al.
Synt. Metals, 123(3), 435-441 (2011)
Preparation and Thermal Stability of Pyrrole and m-Toluidine Copolymer/Montmorillonite Composites
Wang H, et al.
Advanced Materials Research, 915, 780-783 (2014)
Hydrogen-bond-induced clustering in the fragile glass-forming liquid m-toluidine: Experiments and simulations.
Morineau D and Alba-Simionesco C.
J. Chem. Phys. , 109(19), 8494-8503 (1998)
A L Andreeva et al.
Molekuliarnaia genetika, mikrobiologiia i virusologiia, (11)(11), 32-37 (1989-11-01)
Pseudomonas putida strain SU83, harbors the pBS311 plasmid coding for the degradation of biphenyl, 2- and 4-chlorbiphenyl, meta- and paratoluylate. The insertional mutants of the plasmid obtained by the transposon Tn5 insertion were isolated. One of the mutants was used

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service