All Photos(1)
About This Item
Linear Formula:
(CH3CH2OOC)2CHCH(COOCH2CH3)2
CAS Number:
Molecular Weight:
318.32
Beilstein/REAXYS Number:
1805842
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
assay
99%
mp
73-75 °C (lit.)
functional group
ester
SMILES string
CCOC(=O)C(C(C(=O)OCC)C(=O)OCC)C(=O)OCC
InChI
1S/C14H22O8/c1-5-19-11(15)9(12(16)20-6-2)10(13(17)21-7-3)14(18)22-8-4/h9-10H,5-8H2,1-4H3
InChI key
UQSBVZIXVVORQC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Tetraethyl 1,1,2,2-ethanetetracarboxylate was used in the synthesis of double-ring molecules. It was also used in the preparation of tapt and tapt·4HCl·EtOH·0.5H2O.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Dicopper (II) and dicopper (III) complexes with a double-ring octa-aza macrocycle.
Buttafava A, et al.
Journal of the Chemical Society. Chemical Communications, 20, 1166-1167 (1982)
A superoxide dismutase mimic with high activity: crystal structure, solution equilibrium and pulse radiolysis.
J. Chem. Soc., Dalton Trans., 12, 1893-1900 (2000)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service