131172
2-Iminopiperidine hydrochloride
97%
Synonym(s):
2-Amino-3,4,5,6-tetrahydropyridine hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H10N2 · HCl
CAS Number:
Molecular Weight:
134.61
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
assay
97%
form
solid
mp
159-161 °C (lit.)
SMILES string
Cl.N=C1CCCCN1
InChI
1S/C5H10N2.ClH/c6-5-3-1-2-4-7-5;/h1-4H2,(H2,6,7);1H
Inchi Key
ZHDTXTDHBRADLM-UHFFFAOYSA-N
Application
2-Iminopiperidine hydrochloride (2-Amino-3,4,5,6-tetrahydropyridine hydrochloride) was used in the preparation of 2- Benzyl- 1,2,3,4,6,7,8,9-octahydro-11 H-dipyrido[ 1,2-a : 4,3-d]- pyrimidin-11-one.
Reactant for synthesis of:
heteroaromatic compound from alkenes using ketoiodination-cyclization protocols
HIV-1 integrase inhibitor scaffolds
Fluorinated bicyclic heterocycles
Reactant for:
Diels-Alder reactions of heterocyclic azadienes
Condensation reactions
heteroaromatic compound from alkenes using ketoiodination-cyclization protocols
HIV-1 integrase inhibitor scaffolds
Fluorinated bicyclic heterocycles
Reactant for:
Diels-Alder reactions of heterocyclic azadienes
Condensation reactions
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Saturated heterocycles. Part 88. Synthesis of a new ring system: dipyrido [1, 2-a: 4, 3-d] pyrimidin-11-one derivatives.
Huber I, et al.
Journal of the Chemical Society. Perkin Transactions 1, 909-912 (1987)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service