All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C7H13NO · HCl
CAS Number:
Molecular Weight:
163.65
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
assay
97%
form
powder
mp
178-180 °C (lit.)
SMILES string
Cl[H].CCN1CCCC(=O)C1
InChI
1S/C7H13NO.ClH/c1-2-8-5-3-4-7(9)6-8;/h2-6H2,1H3;1H
InChI key
JLBJGYHSFYAUDJ-UHFFFAOYSA-N
Application
1-Ethyl-3-piperidone hydrochloride may be used in chemical synthesis studies.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service