Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

130303

Sigma-Aldrich

1,2-Dimethyl-3-nitrobenzene

97%

Synonym(s):

3-Nitro-o-xylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3NO2
CAS Number:
Molecular Weight:
151.16
Beilstein/REAXYS Number:
2045105
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%
97%

form

liquid

refractive index

n20/D 1.543 (lit.)

bp

245 °C (lit.)

mp

7-9 °C (lit.)

density

1.129 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(c1C)[N+]([O-])=O

InChI

1S/C8H9NO2/c1-6-4-3-5-8(7(6)2)9(10)11/h3-5H,1-2H3

Inchi Key

FVHAWXWFPBPFOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,2-Dimethyl-3-nitrobenzene (3-Nitro-o-xylene) was used to prepare 2-methyl-3-nitrobenzylidene diacetate[1]. It was also used to dissolve sodium-selective ligand[2].

Pictograms

Environment

hcodes

Hazard Classifications

Aquatic Chronic 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 2

flash_point_f

224.6 °F - closed cup

flash_point_c

107 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

J O'Doherty et al.
Science (New York, N.Y.), 203(4387), 1349-1351 (1979-03-30)
The sodium-selective ligand 1,1,1-tris[1(1)-(2(1)-oxa-4(1)-oxo-5(1)-aza-5(1)-methyl)dodecanyl]propane dissolved in 3-nitro-o-xylene containing a small amount of the lipophilic anion tetrachlorophenyl borate was used as a liquid ion-exchanger in sodium-selective microelectrodes. The microelectrodes gave rapid, stable responses that were linear functions of the logarithm of
Oxidation and Claisen condensation products of 3-nitro-o-xylene.
V Askam et al.
Journal of the Chemical Society. Perkin transactions 1, 14, 1935-1939 (1969-01-01)
Brian Roland Larsen et al.
The Journal of physiology, 597(2), 583-597 (2018-10-26)
Neuronal activity induces fluctuation in extracellular space volume, [K+ ]o and pHo , the management of which influences neuronal function The neighbour astrocytes buffer the K+ and pH and swell during the process, causing shrinkage of the extracellular space In
Trine Lisberg Toft-Bertelsen et al.
Glia, 69(1), 28-41 (2020-06-09)
The mammalian brain consists of 80% water, which is continuously shifted between different compartments and cellular structures by mechanisms that are, to a large extent, unresolved. Aquaporin 4 (AQP4) is abundantly expressed in glia and ependymal cells of the mammalian

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service