Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

125741

Sigma-Aldrich

Decahydroquinoline, mixture of cis and trans

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H17N
CAS Number:
Molecular Weight:
139.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.4916 (lit.)

density

0.933 g/mL at 25 °C (lit.)

SMILES string

C1CCC2NCCCC2C1

InChI

1S/C9H17N/c1-2-6-9-8(4-1)5-3-7-10-9/h8-10H,1-7H2

InChI key

POTIYWUALSJREP-UHFFFAOYSA-N

General description

Hydrodenitrogenation of decahydroquinoline has been investigated over Ni-MoS2/γ-Al2O3[1] and NiMo(P)/Al2O3 catalysts[2].

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

154.4 °F - closed cup

flash_point_c

68 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lauren A O'Connell et al.
The Journal of experimental biology, 224(Pt 3) (2021-01-08)
Poison frogs sequester chemical defenses from their diet of leaf litter arthropods for defense against predation. Little is known about the physiological adaptations that confer this unusual bioaccumulation ability. We conducted an alkaloid-feeding experiment with the Diablito poison frog (Oophaga
Ring opening of 1, 2, 3, 4-tetrahydroquinoline and decahydroquinoline on MoS2/γ-Al2O3 and Ni-MoS2/γ-Al2O3.
Gutierrez OY, et al.
J. Catal., 295, 155-168 (2012)
The effect of phosphorus on the HDN reaction of piperidine, decahydroquinoline and ortho-propylaniline over Ni-MoS2/Al2O3 catalysts.
Jian M and Prins R.
Catalysis Letters, 35(3-4), 193-203 (1995)
Stephanie N Caty et al.
The Journal of experimental biology, 222(Pt 12) (2019-05-30)
Poison frogs sequester small molecule lipophilic alkaloids from their diet of leaf litter arthropods for use as chemical defenses against predation. Although the dietary acquisition of chemical defenses in poison frogs is well documented, the physiological mechanisms of alkaloid sequestration
Savithra Jayaraj et al.
Scientific reports, 9(1), 11280-11280 (2019-08-04)
Visible-light mediated aerobic dehydrogenation of N-heterocyclic compounds is a reaction with enormous potential for application. Herein, we report the first complete aerobic dehydrogenation pathway to large-scale production of isoquinolines. The discovery of this visible light photoredox reaction was enabled through

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service