Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

125172

Sigma-Aldrich

4-Chloro-1,3-diaminobenzene

97%

Synonym(s):

4-Chloro-1,3-phenylenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(NH2)2
CAS Number:
Molecular Weight:
142.59
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

assay

97%

mp

87-90 °C (lit.)

functional group

chloro

SMILES string

Nc1ccc(Cl)c(N)c1

InChI

1S/C6H7ClN2/c7-5-2-1-4(8)3-6(5)9/h1-3H,8-9H2

InChI key

ZWUBBMDHSZDNTA-UHFFFAOYSA-N

General description

4-Chloro-1,3-diaminobenzene is dye precursor useful in manufacture of hair dyes[1].

Application

4-Chloro-1,3-diaminobenzene has been used to study the mutagenic effect of phenylenediamines hair chemicals on Salmonella typhimurium TA 102[1].

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charity Mosley-Foreman et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 46(12), 3780-3784 (2008-10-23)
Phenylenediamines (PD) are dye precursors used to manufacture hair dyes. The three PDs, 1,2-,1,3-, and 1,4-PD and three chlorinated PDs, 4-chloro-1,2-PD, 4-chloro-1,3-PD, and 4,5-dichloro-1,2-PD were studied for their mutagenic effect in Salmonella typhimurium TA 102, cytotoxicity in human skin keratinocyte
4-Chloro-ortho-phenylenediamine and 4-chloro-meta-phenylenediamine.
IARC monographs on the evaluation of the carcinogenic risk of chemicals to humans, 27, 81-89 (1982-04-01)
E K Weisburger et al.
Carcinogenesis, 1(6), 495-499 (1980-06-01)
In both male and female Fischer rats, feeding 4-chloro-o-phenylenediamine at 0.5 or 1% in the diet led to a significant increase in hyperplasia, papilloma and carcinoma of the urinary bladder. Neoplastic nodules of the liver and squamous cell papillomas of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service