Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

120421

Sigma-Aldrich

2-(Ethylthio)ethylamine hydrochloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5SCH2CH2NH2 · HCl
CAS Number:
Molecular Weight:
141.66
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

mp

144-146 °C (lit.)

functional group

amine
thioether

SMILES string

Cl.CCSCCN

InChI

1S/C4H11NS.ClH/c1-2-6-4-3-5;/h2-5H2,1H3;1H

InChI key

AZQOTRRSHMSXFJ-UHFFFAOYSA-N

General description

2-(Ethylthio)ethylamine forms complexes with Pd(II) and Pt(II)[1].

Application

2-(Ethylthio)ethylamine hydrochloride was used in the preparation of (syn, syn)-2,7-Bis(N-2-ethylthioethylmethylimino)-1,8-naphthyridine[2]. It was used in the synthesis of N-functionalized bis(phosphino)amine ligands with ether, thioether and pyridyl ethers[3].

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elaboration of 1, 8-naphthyridine-2, 7-dicarboxaldehyde into novel 2, 7-dimethylimine derivatives.
Vu C, et al.
Journal of Heterocyclic Chemistry, 39(4), 829-832 (2002)
Pd (II) and Pt (II) complexes with 2-(ethylthio) ethylamine. Crystal structure of [Pd (2-(ethylthio) ethylamine) Cl2] and [Pd (2-(ethylthio) ethylamine)2](ClO4)2.
Riera X, et al.
Polyhedron, 18(1), 65-73 (1998)
Zhiqiang Weng et al.
Dalton transactions (Cambridge, England : 2003), (32)(32), 3493-3498 (2007-08-08)
Several N-functionalized bis(phosphino)amine ligands with ether, thioether and pyridyl tethers [(R'')2PN(R')P(R'')2=PNP] () have been synthesized. They react with CrCl3(THF)3 in CH2Cl2 to give dinuclear chloro bridged Cr2(micro-Cl)2Cl4(PNP)2 () which converts to the corresponding mononuclear solvento complexes fac-CrCl3(PNP)(NCR) (). The structures

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service