Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

118931

Sigma-Aldrich

3-Bromocinnamic acid, predominantly trans

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4CH=CHCO2H
CAS Number:
Molecular Weight:
227.05
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

mp

177-179 °C (lit.)

solubility

methanol: soluble

SMILES string

OC(=O)\C=C\c1cccc(Br)c1

InChI

1S/C9H7BrO2/c10-8-3-1-2-7(6-8)4-5-9(11)12/h1-6H,(H,11,12)/b5-4+

InChI key

YEMUSDCFQUBPAL-SNAWJCMRSA-N

General description

3-Bromocinnamic acid is the metabolite of cinromide (3-bromo-N-ethylcinnamamide) and can be analysed in plasma by high-performance liquid chromatographic method[1].

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R L DeAngelis et al.
Journal of chromatography, 221(2), 353-360 (1980-12-12)
A quantitative thin-layer chromatography (TLC) procedure is described for the analysis of cinromide (3-bromo-N-ethylcinnamamide) and its two major metabolites, 3-bromocinnamamide and 3-bromocinnamic acid in plasma of the dog. These compounds were recovered from acidified plasma by extraction into benzene with
R J Perchalski et al.
Journal of chromatography, 163(2), 187-193 (1979-06-11)
A high-performance liquid chromatographic method is described for monitoring plasma concentrations of cinromide (3-bromo-N-ethylcinnamamide) and its de-ethylated metabolite. Carbamazepine levels can be easily measured by the same technique. The N-isopropyl analogue of cinromide is used as internal standard, and all

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service