Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

112712

Sigma-Aldrich

Methyl 1-naphthaleneacetate

96%

Synonym(s):

Methyl 1-naphthylacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7CH2CO2CH3
CAS Number:
Molecular Weight:
200.23
Beilstein/REAXYS Number:
1956293
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

96%

refractive index

n20/D 1.597 (lit.)

bp

160-162 °C/5 mmHg (lit.)

density

1.142 g/mL at 25 °C (lit.)

SMILES string

COC(=O)Cc1cccc2ccccc12

InChI

1S/C13H12O2/c1-15-13(14)9-11-7-4-6-10-5-2-3-8-12(10)11/h2-8H,9H2,1H3

InChI key

YGGXZTQSGNFKPJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl 1-naphthaleneacetate undergoes methylenation with Tebbe reagent to form enol ether that is used in the synthesis of nonpeptidic α-helix mimetics.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

228.2 °F - closed cup

flash_point_c

109 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service