Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

112135

Sigma-Aldrich

2,3-Butanedione monoxime

97%

Synonym(s):

BDM, Biacetyl monoxime, Diacetyl monoxime

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C(=NOH)COCH3
CAS Number:
Molecular Weight:
101.10
Beilstein/REAXYS Number:
605582
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

assay

97%

form

solid

bp

185-186 °C (lit.)

mp

75-78 °C (lit.)

SMILES string

CC(=O)\C(C)=N\O

InChI

1S/C4H7NO2/c1-3(5-7)4(2)6/h7H,1-2H3/b5-3+

InChI key

FSEUPUDHEBLWJY-HWKANZROSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Janine E Radford et al.
Protoplasma, 248(1), 205-216 (2010-11-30)
Actin and myosin are components of plasmodesmata, the cytoplasmic channels between plant cells, but their role in regulating these channels is unclear. Here, we investigated the role of myosin in regulating plasmodesmata in a well-studied, simple system comprising single filaments
Stacey L Edwards et al.
Genetics, 201(1), 91-116 (2015-09-12)
The functional integrity of neurons requires the bidirectional active transport of synaptic vesicles (SVs) in axons. The kinesin motor KIF1A transports SVs from somas to stable SV clusters at synapses, while dynein moves them in the opposite direction. However, it
Nicholas M P King et al.
The Journal of general physiology, 137(1), 81-91 (2010-12-29)
A carbon fiber-based cell attachment and force measurement system was used to measure the diastolic stress-sarcomere length (SL) relation of mouse intact cardiomyocytes, before and after the addition of actomyosin inhibitors (2,3-butanedione monoxime [BDM] or blebbistatin). Stress was measured during
Tsung-Ming Shih et al.
Journal of molecular neuroscience : MN, 40(1-2), 63-69 (2009-08-15)
The capability of the tertiary oximes, monoisonitrosoacetone (MINA) and diacetylmonoxime (DAM), to reactivate acetylcholinesterase (AChE) inhibited by sarin (GB) in the blood, brain, and peripheral tissues of guinea pigs was compared with that of the quaternary oximes 2-PAM, HLö7, and
Maozhong Zheng et al.
Planta, 231(4), 779-791 (2009-12-25)
Strategic control of mitochondrial movements and cellular distribution is essential for correct cell function and survival. However, despite being a vital process, mitochondrial movement in plant cells is a poorly documented phenomenon. To investigate the roles of actin filaments and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service