11067
(+)-Aromadendrene
≥97.0% (sum of enantiomers, GC)
Synonym(s):
(1R,2S,8R,11R)-7-Methylene-3,3,11-trimethyltricyclo[6.3.0.02.4]undecane
About This Item
Recommended Products
assay
≥97.0% (sum of enantiomers, GC)
optical activity
[α]20/D +12±1°, neat
refractive index
n20/D 1.497
bp
261-263 °C (lit.)
density
0.912 g/mL at 20 °C (lit.)
storage temp.
2-8°C
SMILES string
[H][C@@]12CC[C@@H](C)[C@@]1([H])[C@@]3([H])[C@@]([H])(CCC2=C)C3(C)C
InChI
1S/C15H24/c1-9-6-8-12-14(15(12,3)4)13-10(2)5-7-11(9)13/h10-14H,1,5-8H2,2-4H3/t10-,11+,12-,13-,14-/m1/s1
InChI key
ITYNGVSTWVVPIC-XVIXHAIJSA-N
General description
Application
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
194.0 °F - closed cup
flash_point_c
90 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service