Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11008

Sigma-Aldrich

Aluminum

grit, ≥97.0% (complexometric)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Al
CAS Number:
Molecular Weight:
26.98
EC Number:
MDL number:
UNSPSC Code:
12141702
PubChem Substance ID:

assay

≥97.0% (complexometric)

form

grit

autoignition temp.

1400 °F

resistivity

2.6548 μΩ-cm

bp

2460 °C (lit.)

mp

660.37 °C (lit.)

density

2.7 g/mL at 25 °C (lit.)

SMILES string

[Al]

InChI

1S/Al

InChI key

XAGFODPZIPBFFR-UHFFFAOYSA-N

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Sol. 1 - Water-react 2

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural features of oxide coatings on aluminum.
Keller F, et al.
Journal of the Electrochemical Society, 100(9), 411-419 (1953)
Aluminum ion diffusion in aluminum oxide.
Paladino AE and Kingery WD.
J. Chem. Phys. , 37(5), 957-962 (1962)
Electrochemistry of Cd (II) in the basic 1-ethyl-3-methylimidazolium chloride/tetrafluoroborate room temperature molten salt.
Chen P-Y and Sun I-W
Electrochimica Acta, 45(19), 3163-3170 (2000)
Thierry Deschamps et al.
Optics express, 21(7), 8382-8392 (2013-04-11)
Using a combination of experimental techniques such as optical absorption, Raman scattering, continuous wave and pulse Electron Spin Resonance (ESR), we characterize a set of γ-irradiated Yb(3+) doped silica glass preforms with different contents of phosphorous and aluminum. We demonstrate
Caitlin Carney et al.
Journal of the American Academy of Dermatology, 69(4), 578-582 (2013-07-17)
Laser treatment has emerged as a novel treatment modality for onychomycosis. We sought to determine thermal response and optical effects of a submillisecond neodymium:yttrium-aluminum-garnet (Nd:YAG) 1064-nm laser on common fungal nail pathogens, and the clinical efficacy and safety of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service