Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

110027

Sigma-Aldrich

Ethyl acetate

≥99.5%

Synonym(s):

EtOAc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
Beilstein/REAXYS Number:
506104
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
assay:
≥99.5%
bp:
76.5-77.5 °C (lit.)
76.5-77.5 °C
vapor pressure:
73 mmHg ( 20 °C)

vapor density

3 (20 °C, vs air)

vapor pressure

73 mmHg ( 20 °C)

assay

≥99.5%

autoignition temp.

801 °F

expl. lim.

2.2-11.5 %, 38 °F

refractive index

n20/D 1.3720 (lit.)
n20/D 1.372

bp

76.5-77.5 °C (lit.)
76.5-77.5 °C

mp

−84 °C (lit.)

density

0.902 g/mL at 25 °C (lit.)

format

neat

SMILES string

CCOC(C)=O

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Respiratory system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

24.8 °F - closed cup

flash_point_c

-4 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Eagleson M.
Concise Encyclopedia Chemistry, 80-80 (1994)
Ethanol dehydrogenation to ethyl acetate by using copper and copper chromite catalysts.
Santacesaria E, et al.
Chemical Engineering Journal, 179, 209-220 (2012)
Heterogeneous Catalytic Hydrogenation of Ethyl Acetate to Produce Ethanol.
Luo M, et al.
Catalysis Today, 57(6-9), 757-761 (2014)
Eagleson M.
Concise Encyclopedia Chemistry, 425-425 (1994)
Towards a green process for bulk-scale synthesis of ethyl acetate: efficient acceptorless dehydrogenation of ethanol.
Martin Nielsen et al.
Angewandte Chemie (International ed. in English), 51(23), 5711-5713 (2012-04-21)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service