Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

109991

Sigma-Aldrich

2,6-Dimethyl-4-heptanone

technical grade, 80%

Synonym(s):

Diisobutyl ketone, Isovalerone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2COCH2CH(CH3)2
CAS Number:
Molecular Weight:
142.24
Beilstein/REAXYS Number:
1743163
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical grade

vapor density

4.9 (vs air)

vapor pressure

1.7 mmHg ( 20 °C)

assay

80%

autoignition temp.

745 °F

expl. lim.

0.8-6.2 %, 100 °F

impurities

<15% 4,6-dimethyl-2-heptanone

refractive index

n20/D 1.412 (lit.)

bp

165-170 °C (lit.)

density

0.808 g/mL at 25 °C (lit.)

SMILES string

CC(C)CC(=O)CC(C)C

InChI

1S/C9H18O/c1-7(2)5-9(10)6-8(3)4/h7-8H,5-6H2,1-4H3

InChI key

PTTPXKJBFFKCEK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

120.2 °F - closed cup

flash_point_c

49 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R F White et al.
Environmental research, 61(1), 117-123 (1993-04-01)
The objective of this investigation was to examine cerebral magnetic resonance imaging (MRI) pathology and functional deficits demonstrated by neuropsychological testing in cases of toxic encephalopathy. Two subjects, occupationally exposed to toxic chemicals, were studied. As part of their neurological
D E Dodd et al.
Toxicology and industrial health, 3(4), 443-457 (1987-12-01)
Four groups, each consisting of 10 male and 10 female Fischer-344 rats, were exposed 6 h/day, 5 days/week, for 9 days to diisobutyl ketone (DIBK) vapor at concentrations of 905, 300, 98, or 0 (control) ppm. An additional 10 rats/sex
[Thermal desorption gas chromatography for determination of diisobutyl ketone in air of working places].
Wei Chen et al.
Zhonghua lao dong wei sheng zhi ye bing za zhi = Zhonghua laodong weisheng zhiyebing zazhi = Chinese journal of industrial hygiene and occupational diseases, 26(2), 110-112 (2008-05-01)
Marte Balchen et al.
Analytica chimica acta, 716, 16-23 (2012-01-31)
A totally new concept based on electrokinetic migration was evaluated for the extraction of three biologically active peptides from human plasma. Angiotensin 2, leu-enkephalin, and endomorphin 1 migrated from a diluted human plasma sample (2 mL, positive electrode), through a
G Milani et al.
Journal of computational chemistry, 28(13), 2203-2215 (2007-04-24)
A simple genetic algorithm for the numerical evaluation of binodal curves in ternary systems polymer-liquid (1)-liquid (2) and polymer (1)-polymer (2)-solvent is presented. The technique exploits a specifically developed restarting technique based on a combined elitist and zooming strategy on

Questions

1–2 of 2 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. What is the difference between 2.6-Dimethyl-4-heptanone, Products 537691 and 109991?

    1 answer
    1. Product No. 537691 is required to have a purity greater than 90%, and Product No. 109991 need only have a purity of around 80%.  Aside from the purity specification, all other specifications are the same.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service