Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

109088

Sigma-Aldrich

Acetic acid

99.8%, anhydrous

Synonym(s):

Glacial acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05
Beilstein/REAXYS Number:
506007
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

anhydrous

vapor density

2.07 (vs air)

vapor pressure

11.4 mmHg ( 20 °C)

assay

99.8%

form

liquid

autoignition temp.

800 °F

expl. lim.

16 %, 92 °F
4 %, 59 °F

refractive index

n20/D 1.371 (lit.)

bp

117-118 °C (lit.)

mp

16.2 °C (lit.)

solubility

water: miscible

density

1.04 g/mL at 25 °C (lit.)

SMILES string

[F2C(F2C)13F3C]C(O)=O

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)

InChI key

QTBSBXVTEAMEQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

102.2 °F - closed cup

flash_point_c

39 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Eagleson M.
Concise Encyclopedia Chemistry, 51-51 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 7-7 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 5-5 (1994)
Takashi Taniguchi et al.
Arthritis & rheumatology (Hoboken, N.J.), 67(2), 517-526 (2014-11-12)
Fli-1, a potential predisposing factor for systemic sclerosis (SSc), is constitutively down-regulated in the lesional skin of patients with SSc by an epigenetic mechanism. To investigate the impact of Fli-1 deficiency on the induction of an SSc phenotype in various
Isabelle Brunet et al.
The Journal of clinical investigation, 124(7), 3230-3240 (2014-06-18)
Autonomic sympathetic nerves innervate peripheral resistance arteries, thereby regulating vascular tone and controlling blood supply to organs. Despite the fundamental importance of blood flow control, how sympathetic arterial innervation develops remains largely unknown. Here, we identified the axon guidance cue

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service