All Photos(1)
About This Item
Linear Formula:
HC6F4NH2
CAS Number:
Molecular Weight:
165.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
assay
97%
bp
158 °C (lit.)
mp
31-32 °C (lit.)
solubility
perchloric acid: soluble
SMILES string
Nc1c(F)c(F)cc(F)c1F
InChI
1S/C6H3F4N/c7-2-1-3(8)5(10)6(11)4(2)9/h1H,11H2
Inchi Key
SPSWJTZNOXMMMV-UHFFFAOYSA-N
General description
2,3,5,6-Tetrafluoroaniline undergoes oxidative electropolymerization to form stable poly(2,3,5,6-tetrafluoroaniline). Poly(2,3,5,6-tetrafluoroaniline) coated on platinum electrodes forms polymer-coated electrocatalytic electrodes, which can be used for direct electricity generation from microbial photosynthetic activity. Poly(2,3,5,6-tetrafluoroaniline) modified Platinum electrodes are stable in microbially aggressive conditions such as sewage or sewage sludge.
Application
2,3,5,6-Tetrafluoroaniline was used to study the formation of electrochromic films of poly(2,3,5,6-tetrafluoroaniline) on ITO substrates from aqueous solutions utilising perchloric acid as dopant.
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
145.4 °F - closed cup
flash_point_c
63 °C - closed cup
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Characterization and electrochromic properties of poly (2, 3, 5, 6-tetrafluoroaniline): Progress towards a transparent conducting polymer.
Astratine L, et al.
Electrochimica Acta, 74, 117-122 (2012)
Fluorinated polyanilines as superior materials for electrocatalytic anodes in bacterial fuel cells.
Niessen J, et al.
Electrochemical Communications, 6(6), 571-575 (2004)
Miriam Rosenbaum et al.
Applied microbiology and biotechnology, 68(6), 753-756 (2005-02-08)
By employing living cells of the green alga Chlamydomonas reinhardtii, we demonstrate the possibility of direct electricity generation from microbial photosynthetic activity. The presented concept is based on an in situ oxidative depletion of hydrogen, photosynthetically produced by C. reinhardtii
Xun Tong et al.
Soft matter, 15(31), 6411-6417 (2019-07-25)
Photoresponsive supramolecular gels have aroused continuous attention because of their extensive applications; however, most studies utilize UV light, which inevitably brings about some health and environmental issues. The halogen bond is an important driving force for constructing supramolecules due to
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service