Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

102547

Sigma-Aldrich

N-Benzyl-N-ethylaniline

99%

Synonym(s):

N-Ethyl-N-phenylbenzylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5N(C2H5)CH2C6H5
CAS Number:
Molecular Weight:
211.30
Beilstein/REAXYS Number:
2099852
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

99%

refractive index

n20/D 1.595 (lit.)

bp

163-164 °C/6 mmHg (lit.)

solubility

alcohol: soluble 1ml in 5.5ml
H2O: insoluble
organic solvents: soluble

density

1.029 g/mL at 25 °C (lit.)

SMILES string

CCN(Cc1ccccc1)c2ccccc2

InChI

1S/C15H17N/c1-2-16(15-11-7-4-8-12-15)13-14-9-5-3-6-10-14/h3-12H,2,13H2,1H3

InChI key

HSZCJVZRHXPCIA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Benzyl-N-ethylaniline can be prepared by reaction of N ethylaniline and benzyl chloride in the presence of phase transfer catalyst[1].

Application

N-Benzyl-N-ethylaniline was used in a study to investigate the electron transfer quenching dynamics of excited perylene and cyanoperylene in various donating solvents[2].

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 2

flash_point_f

314.6 °F - closed cup

flash_point_c

157 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorescence quenching in electron-donating solvents. 2. Solvent dependence and product dynamics.
Morandeira A, et al.
The Journal of Physical Chemistry A, 108(40), 8190-8200 (2004)
Improved Synthesis of N benzyl N ethylaniline [J].
Li YCH, et al.
He Cheng Hua Xue / Chinese Journal of Synthetic Chemistry, 2 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service