Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

09885

Sigma-Aldrich

Ammonium nickel(II) sulfate hexahydrate

purum p.a., ≥98.0% (KT)

Synonym(s):

Nickel ammonium sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2Ni(SO4)2 · 6H2O
CAS Number:
Molecular Weight:
394.99
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

purum p.a.

assay

≥98.0% (KT)

form

powder or crystals

reaction suitability

reagent type: catalyst
core: nickel

mp

85-89 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤500 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤200 mg/kg
Na: ≤100 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

N.N.[Ni++].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].OS(O)(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/2H3N.Ni.2H2O4S.6H2O/c;;;2*1-5(2,3)4;;;;;;/h2*1H3;;2*(H2,1,2,3,4);6*1H2/q;;+2;;;;;;;;/p-2

InChI key

ZZPAXHDZFDUTLY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Diffusivity, density, viscosity, and refractive index of nickel ammonium sulfate aqueous solutions.
Mullin JW and Osman MM.
Journal of Chemical and Engineering Data, 12(4), 516-517 (1967)
The crystal structure of Tutton's salts. II. Magnesium ammonium sulfate hexahydrate and nickel ammonium sulfate hexahydrate.
Montgomery HT and Lingafelter EC.
Acta Crystallographica, 17(11), 1478-1479 (1964)
NAS-benzidine reaction of blood cells-new simple peroxidase staining method using nickel-ammonium sulfate.
T MITSUI et al.
Okajimas folia anatomica Japonica, 23(4-5), 331-336 (1951-08-01)
Ammonium nickel sulfate hexahydrate crystal: a new ultraviolet light filter.
Su G, et al.
Journal of Physics D: Applied Physics, 35(20), 2652-2652 (2002)
J A Moreno-Bravo et al.
Brain structure & function, 219(3), 777-792 (2013-03-16)
Hindbrain rhombomeres in general are differentially specified molecularly by unique combinations of Hox genes with other developmental genes. Rhombomere 1 displays special features, including absence of Hox gene expression. It lies within the hindbrain range of the Engrailed genes (En1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service