Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07404

Sigma-Aldrich

Jone′s reagent

~2% CrO3 basis

Synonym(s):

Chromium trioxide solution in sulfuric acid, Sulfuric acid-chromium trioxide mixture

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

reaction suitability

reagent type: oxidant

concentration

~2% (CrO3)

SMILES string

O=[Cr](=O)=O.OS(O)(=O)=O

InChI

1S/Cr.H2O4S.3O/c;1-5(2,3)4;;;/h;(H2,1,2,3,4);;;

InChI key

HNPXHFOTGAXAJY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Oxidation of oximes to carbonyl compounds[1]
  • Oxidation of benzoins to benzils[2]

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Muta. 1B - Repr. 1B - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT RE 2 Inhalation

target_organs

Cardio-vascular system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxidation of benzoins to benzils with the Jones' reagent supported on graphite
Lou, J.-D.; et al.
Oxidation Communications, 33, 550-554 (2010)
Efficient oxidative conversion of oximes to the corresponding carbonyl compounds with the Jones reagent supported on graphite under heterogeneous conditions
Lou, J.-D.; et al.
Oxidation Communications, 34, 604-609 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service