07086
Tris[4-(heptadecafluorooctyl)phenyl]phosphine
≥85%
Synonym(s):
Tris[4-(perfluorooctyl)phenyl]phosphine
About This Item
assay
≥85%
reaction suitability
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
reaction type: Fluorous synthesis
mp
53-59 °C
functional group
phosphine
SMILES string
FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)c1ccc(cc1)P(c2ccc(cc2)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)c3ccc(cc3)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
InChI
1S/C42H12F51P/c43-19(44,22(49,50)25(55,56)28(61,62)31(67,68)34(73,74)37(79,80)40(85,86)87)13-1-7-16(8-2-13)94(17-9-3-14(4-10-17)20(45,46)23(51,52)26(57,58)29(63,64)32(69,70)35(75,76)38(81,82)41(88,89)90)18-11-5-15(6-12-18)21(47,48)24(53,54)27(59,60)30(65,66)33(71,72)36(77,78)39(83,84)42(91,92)93/h1-12H
InChI key
RTNPUWLBWXIECX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Triarylphosphine selenides
- Perfluoroalkylated arylphosphines
Reactant for:
- Suzuki coupling
- Stille coupling
Legal Information
signalword
Danger
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1
target_organs
Liver
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
>230.0 °F
flash_point_c
> 110 °C
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service