Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

06694

Sigma-Aldrich

Tris(3,3,4,4,5,5,6,6,6-nonafluorohexyl)tin hydride

≥90%

Synonym(s):

Tris(1H,1H,2H,2H-perfluorohexyl)tin hydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H13F27Sn
CAS Number:
Molecular Weight:
860.96
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

assay

≥90%

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)CC[SnH](CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)F)CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/3C6H4F9.Sn.H/c3*1-2-3(7,8)4(9,10)5(11,12)6(13,14)15;;/h3*1-2H2;;

InChI key

IXGCTPQSODDMDJ-UHFFFAOYSA-N

Legal Information

Product of Fluorous Technologies, Inc. Protected by U.S. patents 6,156,896; 5,859,247; and 5,777,121

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service