Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

00454

Sigma-Aldrich

Diisopropyl(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane

≥95%

Synonym(s):

Diisopropyl-(1H,1H,2H,2H-perfluorooctyl)silane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19F13Si
CAS Number:
Molecular Weight:
462.37
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

assay

≥95%

SMILES string

[H][Si](CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)(C(C)C)C(C)C

InChI

1S/C14H19F13Si/c1-7(2)28(8(3)4)6-5-9(15,16)10(17,18)11(19,20)12(21,22)13(23,24)14(25,26)27/h7-8,28H,5-6H2,1-4H3

InChI key

VIGKXSZWCIUYJE-UHFFFAOYSA-N

Legal Information

Product of Fluorous Technologies, Inc. Use of this compound may be protected by U.S. patents 6,156,896; 5,859,247; 5,777,121 and 6,673,539.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service