MilliporeSigma
Sustainable Detergents for Virus Inactivation and Cell Lysis
Our new offer of Emprove®detergents for a sustainable Triton™ X-100 detergent replacement for biomanufacturing
*Triton™ is a trademark of The Dow Chemical Company
Detergents Use In Bioprocessing
Detergents, such as zwitterionic non-ionic surfactants, are used in biomanufacturing processes for a variety of applications including viral inactivation and cell lysis.
Deviron® C16
Deviron® 13-S9
Looking For A Triton™ X-100 Alternative?
Triton™ X-100 (4-tert-octylphenol polyethoxylate), a non-ionic detergent, is commonly used for virus inactivation and cell lysis as it disrupts the lipid membrane of cells and enveloped viruses.
Triton™ X-100 is the main detergent templated in biomanufacturing processes, but as of January 2021, its unauthorized use has been prohibited in the European Union by the European Commission, due to its listing to REACH (Registration, Evaluation, Authorization and Restriction of Chemicals).
In response to the global implications of restrictions on Triton™ X-100 detergent, we have developed the Deviron® portfolio of sustainable detergents.
The Deviron® Detergent Portfolio as a Sustainable Triton™ X-100 Alternative
Our Deviron® portfolio of sustainable detergents should be considered as a primary alternative to Triton™ X-100 by all biologics manufacturers.
To address the need for alternatives to Triton™ X-100 that do not require a REACH authorization, we have launched the Deviron® C16 detergent and the Deviron® 13-S9 detergent.
Deviron® C16 Emprove® Evolve Detergent
Deviron® 13-S9 Emprove® Expert Detergent
Triton™ X-100 Emprove® Expert Detergent
Product code
108693
108694
108643
Surfactant type
Zwitterionic (pI 8.9)​
Non-ionic​
Non-ionic​
Chemical name
N,N-Dimethyltetradecylamin-N-oxid
Alcohols, C11-15-secondary, ethoxylated
4-(1,1,3,3-tetramethylbutyl)phenol ethoxylated
CAS number
3332-27-2
68131-40-8
9036-19-5
Critical Micelle Concentration (CMC)
0.002-0.003 % wt​ (24 °C)
0.005 %​ wt (24 °C)
0.013 % wt (24 °C)
Biodegradibility (Readily biodegradable: >60 % within 28 days) OECD 301B
Readily biodegradable
88 % degradation
Readily biodegradable
74 % degradation
Substance of Very High Concern as defined by the REACH1
Virus inactivation efficiency (LRV ≥ 4).
(Detergent ≥0.5 % wt
T=15-25 °C)
>5 LRV
>5 LRV
Benchmark
Cell lysis efficiency for viral vector processing
Better or comparable to benchmark
Better or comparable to benchmark
Benchmark
Endotoxins removal for plasmid manufacturing:
Natrix® Q chromatography membrane/ detergents added to wash buffer supplement
99.997 % removal
99.969 % removal
Benchmark
Emprove® & MQ level
Emprove® Evolve, MQ400, (ISO9001 or equivalent)
Emprove® Expert, MQ500, (IPEC-PQG GMP)​
Emprove® Expert, MQ500, (IPEC-PQG GMP)​
Composition
30 % wt. water solution
Pure substance (100 %)
Pure substance (100 %)
Available formats
1L, 2.5 L, 25 L
1 L, 2.5 L, 25 L
1 L, 2.5 L, 25 L, 190 L
Customized Certificate of Analysis options
Available
Available
Available
Detection method
HPLC-ELSD method available
HPLC-ELSD method available
*Benchmark: Triton™ X-100 detergent
KEY CONSIDERATIONS FOR DETERGENT SELECTION IN BIOPROCESSING
Several factors should be considered when evaluating detergents in biomanufacturing.
These include:
  • Biodegradability (REACH* Compliance) and lack of harmful degradation products
  • Viral inactivation (5 Log Reduction Value)
  • Cell lysis efficiency
  • Minimal impact on product functionality or quality
  • Lack of process interference
  • Effective removal by downstream operations
  • Availability of sensitive detection method
  • Manufacturability of high volumes as IPEC-PQG GMP quality material
  • Impurity profile compatible with parenteral usage
*Registration, Evaluation, Authorisation and Restriction of Chemicals
Virus Inactivation for CHO Cell Harvest and Human Plasma Protein Purification
Viral safety is a major concern for biotherapeutic manufacturers.
  • Cell-based processes may produce endogenous retroviral particles, and adventitious viruses can be introduced from contaminated source materials or during the manufacturing process.
  • Human plasma-derived products are at risk of containing viruses, despite extensive screening of donation material.
Detergent-mediated viral inactivation is widely used in multiple biotherapeutic production processes as part of an overall virus safety strategy.
Our Deviron® portfolio shows efficient viral inactivation for both matrices with different model viruses and operating conditions.
Virus inactivation was assessed in mAb-containing CHO clarified harvest and human plasma matrices with XMuLV (Xenotropic Murine Leukemia Virus).
  • The standard viral inactivation practice is described in the ASTM E3042-16.
  • mAb processes >0.5%, no solvent and incubation time >60 min.
  • Plasma processes typically utilize 1% detergent /0.3% solvent TnBP and longer incubation time (4-6 h).
Deviron® C16 and Deviron® 13-S9 demonstrate effective viral inactivation (LRV >5) in all conditions tested. Additional data with PRV and BVDV viruses are available. 
Viral inactivation (XMuLV) with Deviron® C16 in CHO clarified harvest at 22°C compared to Triton™ X-100
Deviron® C16 in CHO clarified harvest at 22°C
Viral inactivation (XMuLV) with Deviron® 13-S9 in CHO clarified harvest at 15° compared to Triton™ X-100
Deviron ® 13-S9 in CHO clarified harvest at 15°C
Viral inactivation (XMuLV) with Deviron® C16 in neat plasma with 0.3% TnBP at 22°C compared to Triton™ X-100
Deviron ® C16 in neat plasma with 0.3% TnBP at 22°C
Viral inactivation (XMuLV) with Deviron® 13-S9 in cryo-poor plasma with 0.3% TnBO at 22° compared to Triton™ X-100
Deviron ® 13-S9 in cyro-poor plasma with 0.3% TnBP at 22°C compared to Triton™ X-100
Viral inactivation (XMuLV) with Deviron® C16 in CHO clarified harvest at pH 5.5 and pH 8.5 at 22°
Deviron ® C16 in CHO clarified harvest at pH 5.5 and pH 8.5 at 22°C
Viral inactivation (XMuLV) with Deviron® 13-S9 in IgG at pH 6 and pH 8 at 15°
Deviron ® 13-S9 in IgG at pH 6 and pH 8 at 15°C
Cell Lysis for AAV Manufacturing
For biomanufacturing production of viral vectors used as therapeutics, a lysis step is needed to release the viral particles of interest. For Adeno Associated Virus (AAV) vectors used for gene therapies, host cells (HEK293 or Sf9 cells) used for production of viral vectors are often lysed using detergents like Triton™ X-100 or Polysorbates 20/80.
We introduce two new cell lysis options with our Deviron® detergent portfolio.
Cell lysis efficiency was assessed with HEK293 cells. After one day of cultivation, the cells were transfected with the selected plasmid polyethylenimine (PEl) complexation. After 72 hours of cultivation, the cells were harvested and lysate prepared according to the following procedure:
  • Detergent concentration: 0.5% wt/vol
  • Detergent: Tween™ 20 and Triton™ X-100 (benchmarks), Deviron® C16 and Deviron® 13-S9
  • Nuclease: 25 U/mL Benzonase® with 2 mM MgCl2
  • Lysis time: 2 hr
  • Temperature: 37 °C
Following incubation, each sample was counted with ViCell and total cell count and viability determined.
Cell viability for HEK293 Adherent cells in AAV2 production
HEK293 Adherent cells – AAV2
Cell viability for HEK293 suspension cells in AAV5 production
HEK293 suspension cells – AAV5
THE EMPROVE® PROGRAM FOR DETERGENTS
Roadmap for Choosing Fit-For-Purpose Raw Materials
As a supplier of thousands of raw materials for biologics manufacturing, we know the importance of quality and consistency. Our Emprove® program is designed to provide customers with the transparency and information needed to select fit-for-purpose raw materials and access to detailed documentation to meet regulatory requirements. 
DEVIRON® C16 Emprove® Evolve 
Provides detailed and transparent supply chain information and documentation to support risk assessments for critical raw materials used in manufacturing processes. Non-GMP but Manufactured utilizing GMP concepts and elements; ISO9001 or equivalent.
DEVIRON® 13-S9 Emprove® Expert
Addresses higher risk applications, where the lowest microbiological and endotoxin levels are of utmost importance. Released with IPEC-PQG GMP.
Related Product Resources
Request Information