MilliporeSigma
HomeProductsIndustrial MicrobiologyEnvironmental Monitoring and Aseptic Process Simulation
Environmental Monitoring and Aseptic Process Simulation
Patient safety is crucial, thus pharmaceutical products are manufactured under stringent controls. Microbial monitoring plays a vital role in GMP regulatory compliance, demonstrating that manufacturing processes, particularly in aseptic production, are well-managed.
Explore our comprehensive range of GMP-compliant solutions for microbial monitoring in aseptic pharmaceutical manufacturing, which includes monitoring of surfaces, personnel, and air. We also offer tailored solutions for isolators and RABS to enhance your workflow.
Additionally, we provide both granulated and ready-to-use culture media for media fills and aseptic process simulations, supporting classical environmental monitoring.
Products
Sort by Relevance
Showing 1-20 of 172
Change View
Showing 1-20 of 172
Compare
Product Number
Product Name
Product Description
Pricing
MICROBIAL AIR SAMPLERS
Our air samplers are based on different principles, enabling technology selection to suit your needs. We offer products specifically designed for explosion hazard areas, cleanrooms and isolators, and compressed gas monitoring. Our easy-to-use monitoring systems are designed for many environmental monitoring needs.
  • The MAS-100 NT® and MAS-100 NT Ex® portable  air samplers with or without HEPA exhaust filter provide excellent accuracy, precision and sampling efficiency. They are designed for highly critical areas, such as cleanrooms grade A
  • The MAS-100 VF® active air sampler is designed for controlled environments. It is compact, easy to handle, and perfect for monitoring environment quality.
  • Easily and automatically detect microbial contamination in gases with the MAS-100 Atmos® compressed gas sampler, which is simple, safe, and compliant with 21 CFR Part 11.
CONTACT PLATES, SETTLE PLATES, SLIDES AND SWABS
We provide a wide selection of plates tailored for environmental monitoring in isolators and critical cleanrooms, including the non-lockable ICR and lockable ICRplus Contact and Settle Plates with TSA + LTHTh.
  • ICR Swabs are designed for absence-presence testing on dry surfaces in isolators and cleanrooms that are challenging to access for surface and personnel monitoring.
  • ICR Contact plates are designed to minimize the risk of contamination as they are produced in clean rooms. The lockable ICRplus/RTplus versions can be safely transported for incubation under aerobic, microaerophilic, or anaerobic conditions.
  • Hycon® Contact Slides are intended for monitoring flat and curved surfaces and personnel. They are available in standard single packaging or a double-bagged, gamma-irradiated format.
  • ICR and ICRplus settle plates feature a high filling volume of 30 mL in 90 mm plates to reduce water loss during air monitoring. They are offered in animal-free formulations to minimize the risk of BSE/TSE contamination.
ISOLATORS AND RABS SOLUTIONS
Isolators and RABS are designed to eliminate human contact during controlled manufacturing.
  • MAS-100 Iso NT® & MAS-100 Iso MH® – fulfill the specific requirements to monitor microbial contamination in critical areas.
  • IsoBag® rapid transfer bag – enable safe remote microbial air sampling and rapid culture media transfer into isolators. The IsoBag® solution expedites environmental monitoring in aseptic production isolators by providing ready-to-use gamma-irradiated contact or settle plates for immediate use.
CULTURE MEDIA FOR ASEPTIC PROCESS SIMULATION
Our irradiated, triple-wrapped granulated media or ready-to-use media alleviate concerns about compromising validated processes during media fill trials.
  • Granulated culture media, carefully formulated, enables pharmaceutical manufacturers to conduct efficient media fill trials with minimal risks.
  • Ready-to-use culture media, such as Tryptic Soy Broth (TSB) and Vegetable Peptone Broth (VPB), undergo sterilization by autoclave and a two-step pre-filtration process, ensuring their quality.
Related Resources
Page 1 of 3