MilliporeSigma
Titration Reagents
The image depicts a white bottle of Titripac liquid solution alongside its packaging box. The bottle has a black cap and is partially wrapped with a label containing text and the brand’s logo. The packaging box is white with green accents, displaying information including the product name, usage instructions, and a QR code. Both the bottle and box are placed against a light-colored, plain background. The box has a cutout handle for easy carrying.
Titration or titrimetry is a quantitative analytical technique used to determine the exact concentration of an unknown compound or the purity of a substance. Sophisticated and accurate analyses require precisely adjusted and produced volumetric solutions. Our broad portfolio of volumetric solutions in different concentrations and packaging has a solution for every application. Especially our Titripur® Volumetric Solutions are the quality standard for titration, they are ready to use, analyzed in our ISO/IEC 17025 accredited calibration laboratory. 
View Popular Filters:
Products
Sort by Relevance
Showing 1-20 of 168
Change View
Showing 1-20 of 168
Compare
Product Number
Product Name
Product Description
Pricing
Titripur® Volumetric Solutions
Our Titripur® products include ready-to-use volumetric solutions for precise and reliable titration results. All Titripur® solutions are traceable to primary reference materials from the National Institute of Standards and Technology (NIST) and analyzed by our ISO/IEC 17025 accredited laboratory. The Quality Control Laboratory at Merck KGaA, Darmstadt, Germany is accredited by DAkkS (Deutsche Akkreditierungsstelle- German Accreditation Service) as calibration laboratory D-K-15185-01-00 according to DIN EN ISO/IEC 17025. Our Titripur® portfolio offers also a range of solutions which are produced and analyzed in adherence to the reagents chapter of the European and United States Pharmacopeia for pharmaceuticals and raw materials analysis.
Titripac® Packaging for Volumetric Solutions
The Titripac® system is an innovative packaging system for our ready-to-use Titripur® volumetric solutions. The hermetically sealed Titripac® packaging prevents solution contamination caused by air, carbon dioxide, or microorganisms through both storage and use. With its integrated tap, solutions can be withdrawn and directly connected to a titrator using an adapter. It is environment friendly and reduces packaging waste; the carton can be recycled with paper waste and the internal liner can be easily folded together and disposed separately.
Titrisol® Volumetric Concentrates
Titrisol® concentrates provide flexiblity to produce different solution concentrations, allowing users to dilute to their required titrant concentration.
SmartChemicals for Digitalized Titration
Supelco® SmartTitrants and Supelco® SmartStandards provide instant and seamless data transfer via RFID tags. This smart titration technology stores relevant certificate data in the embedded RFID tag on the labels of our Titripur® volumetric solutions, Certipur® volumetric standards, and Aquastar® Karl Fischer titrants and standards. A simple touch on the tag of our SmartChemicals enables the transfer of product name, article number, batch number, concentration, date of release, shelf life, and initial opening date.
Related Resources
Related Webinars
A digital illustration of a laptop displaying a simplistic human figure next to a pink rectangle, all set against a yellow background.
Tips & Tricks to Improve Your Titration Process
Learn what parameters could influence your titration process and how to improve your daily workflow. You will also get an insight into our latest innovation of seamless data transfer.
A digital illustration of a laptop displaying a simplistic human figure next to a pink rectangle, all set against a yellow background.
Volumetric Titration: A Step-by-Step Guide to Correct Results
In this expert webinar, we explore the basics of volumetric titration and present tips and advice to help you make sure that your titration results are always precise and reproducible.