Merck
  • Impact of herbicide Ametryn on microbial communities in mixed liquor of a membrane bioreactor (MBR).
Impact of herbicide Ametryn on microbial communities in mixed liquor of a membrane bioreactor (MBR).
Bioresource technology (2012-01-11)
Dimuth Navaratna, Jennifer Elliman, Alanna Cooper, Li Shu, Kanagaratnam Baskaran, Veeriah Jegatheesan
ABSTRACT
Ametryn, which is a second generation herbicide, was introduced to a lab-scale MBR at a concentration of 1mg/L and a 20-40% removal was observed at HRT ranging from 7.8 to 15.6h for an average influent Ametryn concentration of 0.8 mg/L. Components of EPS (protein and carbohydrates) increased in the bioreactor and the observed biomass production reduced after the addition of Ametryn. In a batch study, GAC was added to MBR mixed liquor and removal of Ametryn via biodegradation and adsorption were measured. Five common bacterial colony types (Gram negative and positive bacilli and Gram negative cocci) were identified and three of these were resistant to Ametryn up to 5mg/L. GAC was found to be a very effective Ametryn adsorption medium and in some occasions Ametryn may have acted as a nutrient source for bacteria.
Product Description
Supelco
Ametryn, PESTANAL®, analytical standard
Pricing and availability is not currently available.