Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of disk
manufacturer/tradename
NIST®
technique(s)
XRF: suitable
optical emission spectroscopy (OES): suitable
format
matrix material
Looking for similar products? Visit Product Comparison Guide
General description
This Standard Reference Material (SRM) is designed for use in optical emission, x-ray fluorescence and spectrochemical methods of analysis. For more information, please refer to the SDS and COA.
SRM 1113_cert
SRM 1113_SDS
SRM 1113_cert
SRM 1113_SDS
Other Notes
Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Copper (Cu), Zinc (Zn), Tin (Sn), Nickel (Ni), Iron (Fe), Lead (Pb), Phosphorus(P)
See certificate for values and more details at nist.gov/SRM.
Copper (Cu), Zinc (Zn), Tin (Sn), Nickel (Ni), Iron (Fe), Lead (Pb), Phosphorus(P)
See certificate for values and more details at nist.gov/SRM.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
related product
Product No.
Description
Pricing
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service