Merck
  • Preclinical evaluation of norgestimate, a progestin with minimal androgenic activity.
Preclinical evaluation of norgestimate, a progestin with minimal androgenic activity.
American journal of obstetrics and gynecology (1992-10-01)
A Phillips, D W Hahn, J L McGuire
ABSTRACT
Norgestimate is a novel progestin with highly selective progestational activity and minimal androgenicity. In rabbits, norgestimate binds to uterine progestin receptors, stimulates the endometrium, and inhibits ovulation. Norgestimate acts directly on target organs, stimulating rabbit endometrium when injected into the uterine cavity and inhibiting luteinizing hormone-releasing hormone-stimulated luteinizing hormone release in dispersed rat pituitary cells in culture. Norgestimate has no estrogenic activity, and like other progestins, it suppresses the action of estrogen. Unlike some other progestins, it is relatively free of androgenic activity. Norgestimate and its 17-deacetylated metabolite demonstrate very poor affinity for androgen receptors compared with levonorgestrel and gestodene and do not exhibit androgenic activity when measured as the stimulation of prostatic growth in immature rats. Norgestimate's lack of affinity for human sex hormone-binding globulin is further evidence of its minimal androgenicity.
Product Description
Norgestimate for system suitability, European Pharmacopoeia (EP) Reference Standard
SKU
Pack Size
Availability
Price
Quantity
Norgestimate, European Pharmacopoeia (EP) Reference Standard