EP0030128931
Eppendorf® Twin tec PCR plates, 384 wells
colorless plate, conical bottom colorless wells, pkg of 300 ea
All Photos(1)
About This Item
UNSPSC Code:
41106300
NACRES:
NB.15
material:
colorless plate
conical bottom colorless wells
conical bottom colorless wells
size:
384 wells
Recommended Products
material
colorless plate
conical bottom colorless wells
packaging
pkg of 300 ea
manufacturer/tradename
Eppendorf® 0030128931
technique(s)
PCR: suitable
size
384 wells
well maximum volume
40 μm
General description
The Eppendorf twin.tec® PCR plate combines extremely thin-walled polypropylene wells for optimal heat transfer to the sample, with robust polycarbonate frames for high rigidity. With raised well rims, twin.tec® plates enable highly effective sealing to reduce the risk of cross-contamination. With twin.tec® PCR plates you’ll have maximum confidence in your results.
Application
Eppendorf® Twin tec PCR plates, 384 wells are suitable for use in:
- Standard PCR
- Real-time PCR
- High throughput PCR
- Small volume sample handling
Features and Benefits
- Batch -tested and certified to be DNA, DNase, RNase, and PCR inhibitors free (PCR clean).
- Specially optimized rim is suitable for barcodes and automated work processes.
- The plates are stackable.
- They adhere to the recommended SBS footprint standards.
- These plates are ideal for a 384-well thermocycler, well-suited for the Eppendorf Mastercycler pro-384.
- Maximum volume per well is 40 µL.
- Upon request, the plates can be provided with barcodes, but this option is available only for skirted and semi-skirted variants.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)