Skip to Content
Merck
All Photos(1)

Key Documents

HG0000C2

Millipore

MacConkey Agar

salt yes, crystal violet yes, pH 7.1±0.2 (25 °C, after autoclaving)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105

composition

Agar, 13.5 g/L
Bile Salts No 3, 1.5 g/L
Crystal Violet, 0.001 g/L
crystal violet: yes
Lactose, 10.0 g/L
Neutral Red, 0.03 g/L
Peptone, 20.0 g/L
salt: yes
Sodium chloride, 5.0 g/L

packaging

pkg of 12 × 500 g (plastic bottles)

pH

7.1±0.2 (25 °C, after autoclaving)

storage temp.

15-25°C

Application

A selective agar suitable for the isolation of Salmonella, Shigella and coliform bacteria from foods, urine, faeces, waste water, etc.

Preparation Note

Suspend 50g in 1 litre demineralised water. Boil whilst stirring until completely dissolved. Autoclave at 121°C for 15 minutes. Cool to 45 -50°C. Mix well and pour plates.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service