Skip to Content
Merck
All Photos(1)

Key Documents

SAAR2233500

Ethanol Rectified

rectified, 96%, uniLAB

Synonym(s):

Ethanol

Sign Into View Organizational & Contract Pricing

Select a Size

SAAR2233500LP
R 1 276,94

R 1 276,94

List PriceR 1 303,00

Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
SAAR2233500LP
R 1 276,94

About This Item

Empirical Formula (Hill Notation):
C2H6O
Molecular Weight:
46.07
UNSPSC Code:
15101802

R 1 276,94

List PriceR 1 303,00

Please contact Customer Service for Availability

Request a Bulk Order

product line

uniLAB

Assay

96%

form

liquid

packaging

poly bottle of 1 × 2.5 L (SAAR2233500LP)

dilution

(for laboratory use only)

density

0.81 g/mL

format

neat

Looking for similar products? Visit Product Comparison Guide

Application

For laboratory use only.
chemically pure

Legal Information

uniLAB is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service