Skip to Content
Merck
All Photos(1)

Documents

66979-U

Supelco

BIOshell A160 Peptide CN (2.7 μm) HPLC Columns

L × I.D. 0.5 cm × 4.6 mm, HPLC Guard Cartridge, pkg of 3 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

product name

BIOshell A160 Peptide CN, 2.7 μm Guard Cartridge, 2.7 μm particle size, L × I.D. 0.5 cm × 4.6 mm, pkg of 3 ea

material

stainless steel hardware (column)

Agency

suitable for USP L10

description

Shell thickness (0.5 μm)
Solid Core (1.7 μm)

product line

BIOshell

feature

endcapped

packaging

pkg of 3 ea

extent of labeling

2.2% carbon loading

parameter

600 bar max. pressure
90 °C max. temp.

technique(s)

HPLC: suitable
LC/MS: suitable
UHPLC-MS: suitable
UHPLC: suitable

L × I.D.

0.5 cm × 4.6 mm

surface area

90 m2/g

surface coverage

2.2 μmol/m2

matrix

spherical silica particle platform
superficially porous particle

matrix active group

cyano phase

particle size

2.7 μm

pore size

160 Å

operating pH range

1-8

separation technique

hydrophilic interaction (HILIC)
normal phase
reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

BIOshell Guard Columns provide physical (filtration) and chemical protection for costly analytical columns without compromising the very high performance of BIOshell columns. These BIOshell guard columns are capable of continuous use at pressures up to 9000 psi (600 bar) with only hand-tightening. Guard cartridges are easily replaced without removing the guard column holder from the flow path. The cartridge is packed with BIOshell Fused-Core particles. Order the guard column holder (66841-U) separately.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service