Skip to Content
Merck
All Photos(1)

Documents

57659

Union

bore 0.020 in., 10-32 thread

Synonym(s):

Upchurch PEEK Fingertight Union

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

bore

0.020 in.

fitting

10-32 thread

Looking for similar products? Visit Product Comparison Guide

General description

The PEEK zero dead volume unions (bore 0.020 in., thread 10-32) need to be operated at 5000 ψ (352 kg/cm2). They include Fingertight III nuts and ferrules.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service