Skip to Content
Merck
All Photos(1)

Key Documents

40141-U

Supelco

DBA Isocyanate Standard Mix

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
Pricing and availability is not currently available.

feature

standard type calibration

packaging

ampule of 6 × 1 mL

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Isocyanic acid-di-n-butylamine (ICA-DBA) 1 μg/mL
    Ethyl isocyanate-di-n-butylamine (EIC-DBA) 1 μg/mL
    Hexamethylene diisocyanate-2(di-n-butylamine) (HDI-2(DBA) 1 μg/mL
    Isophorone isocyanate-2(di-n-butyl amine) isomer 1 (IPDI-2(DBA) 1 μg/mL
    Isophorone isocyanate-2(di-n-butyl amine) isomer 2 (IPDI2-2(DBA) .4 μg/mL
    4,4′-Methylenediphenyl diisocyanate-2(di-n-butylamine) (4,4′-MDI-2(DBA) 1 μg/mL
    Methyl isocyanate-di-n-butylamine (MIC-DBA) 1 μg/mL
    Phenyl isocyanate-di-n-butylamine (PhI-DBA) 1 μg/mL
    Propyl isocyanate-di-n-butylamine (PIC-DBA) 1 μg/mL
    2,4-Toluene diisocyanate-di-n-butylamine (2,4-TDI-2(DBA) 1 μg/mL
    2,6-Toluene diisocyanate-2(di-n-butylamine) (2,6-TDI-2(DBA) 1 μg/mL
    See All (11)

Application

Product No.
Description
Pricing

sample collection

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chromatograms

application for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service