Skip to Content
Merck
All Photos(1)

Documents

Z740368

Urisafe® urine collection container

size 24 hour, volume 4 L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

packaging

pack of 30 ea

manufacturer/tradename

Simport B350-4L

size

24 hour

volume

4 L
40000 mL

Looking for similar products? Visit Product Comparison Guide

General description

The patented URISAFE Urine Collection Containers are USER FRIENDLY to both patients and laboratory personnel. Two sizes are available: 3 and 4 liters­.

1 The unique snap valve pour spout is easily popped open and offers dripless pouring
2 Specially designed leakproof screw cap with liner for safer transport
3 Large centrally located handle for good balance and ease of manipulation
4 Metal free and resistant to hydrochloric acid
5 Sample volumes can be read in 100 ml increments

Features and Benefits

The innovative cap:
  • Snap valve pouring spout incorporated
  • Leakproof: a cap liner ensures safe sample transport

The unique snap valve pour spout:
  • Easily popped open or pushed shut
  • Offers dripless pouring
  • Controlled flow rate for better handling of poured volumes
  • Reduced risks of aerosol contamination when pouring
  • Eliminates splashing and exposure to hazardous body fluids

Low form design:
  • Convenient refrigerator storage with less wasted space
  • Anatomically designed for ease of patient use

Large central handle:
  • Can be gripped comfortably with three fingers
  • Permits pouring off of samples with ease and reduces fatigue

Chemically resistant:
  • Metal, latex, zinc and fluorescence free
  • Resistant to hydrochloric acid.
  • Can be subjected to freezing, thawing and EtO gas sterilisation without causing changes in materials or physical appearance

Graduated vertically and horizontally:
  • Graduations are easy to read
  • Place container upright and read in 100 ml increments
  • Can be gamma sterilized

Legal Information

Urisafe is a registered trademark of Simport

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service