Skip to Content
Merck
All Photos(1)

Key Documents

T2900

Sigma-Aldrich

Thr-Arg

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H21N5O4
CAS Number:
Molecular Weight:
275.30
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

composition

Peptide content, ≥65%

storage temp.

−20°C

SMILES string

OS(O)(=O)=O.C[C@@H](O)[C@H](N)C(=O)N[C@@H](CCCNC(N)=N)C(O)=O.C[C@@H](O)[C@H](N)C(=O)N[C@@H](CCCNC(N)=N)C(O)=O

InChI

1S/2C10H21N5O4.H2O4S/c2*1-5(16)7(11)8(17)15-6(9(18)19)3-2-4-14-10(12)13;1-5(2,3)4/h2*5-7,16H,2-4,11H2,1H3,(H,15,17)(H,18,19)(H4,12,13,14);(H2,1,2,3,4)/t2*5-,6+,7+;/m11./s1

InChI key

PPZXSKKWYLGKOO-DHRNAUBXSA-N


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Himani Agrawal et al.
Food research international (Ottawa, Ont.), 120, 697-707 (2019-04-20)
Antioxidant peptides were successfully identified from a finger millet protein hydrolysate. The trypsin hydrolysate showed a higher degree of hydrolysis (17.47 ± 0.63%) than the pepsin hydrolysate, was further fractionated by ultrafiltration membrane [<3 kDa (UF3), 3-10 kDa (UF2) and > 10 kDa (UF1)]. UF3 with higher
Daniel J Wolter et al.
The Journal of antimicrobial chemotherapy, 76(3), 616-625 (2020-12-02)
Bacteria adapt to survive and grow in different environments. Genetic mutations that promote bacterial survival under harsh conditions can also restrict growth. The causes and consequences of these adaptations have important implications for diagnosis, pathogenesis, and therapy. We describe the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service