SRP5102
Mortalin (47-end), GST tagged human
recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
CSA, GRP75, HSPA9, HSPA9B, MGC4500, MOT, MOT2, MTHSP75, PBP74, mot-2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
biological source
human
recombinant
expressed in baculovirus infected Sf9 cells
Assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
mol wt
~97 kDa
NCBI accession no.
application(s)
cell analysis
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... HSPA9(3313)
General description
Mortalin is a mitochondrial chaperone and a member of the heat shock protein 70 family that is constitutively expressed in cells. Mortalin plays a central role in mitochondrial biogenesis through its capacity to direct the import of nuclear-encoded proteins into the mitochondria. Mortalin plays a role in the control of cell proliferation and elevated levels of mortalin have correlated with malignant transformation and poor cancer prognosis. Mortalin can support cancer cell resistance to complement-dependent cytotoxicity. Mortalin expression was decreased in the mitochondrial fraction of neurons from the substantia nigra of Parkinson disease patients.
Physical form
Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.
Preparation Note
after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of neural transmission (Vienna, Austria : 1996), 116(10), 1289-1293 (2009-08-07)
Mortalin is a mitochondrial chaperone of the heat shock protein 70 family. Mortalin plays a central role in mitochondrial biogenesis through its capacity to direct the import of nuclear-encoded proteins into the mitochondria. As mitochondrial dysfunction has been involved in
International journal of cancer, 126(6), 1428-1435 (2009-09-10)
Mortalin, the mitochondrial hsp70, is a vital constitutively expressed heat shock protein. Its elevated expression has been correlated with malignant transformation and poor cancer prognosis. Cancer cells exhibit increased resistance to complement-dependent cytotoxicity, partly due to their capacity to eliminate
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service