Skip to Content
Merck
All Photos(2)

Documents

SAB2104260

Sigma-Aldrich

Anti-EBF2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-COE2, Anti-EBF-2, Anti-FLJ11500, Anti-O/E-3, Anti-OE-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

63 kDa

species reactivity

bovine, horse, human, rabbit, mouse, rat, guinea pig, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EBF2(64641)

Immunogen

Synthetic peptide directed towards the middle region of human EBF2

Biochem/physiol Actions

EBF2 belongs to the conserved Olf/EBF family of helix-loop-helix transcription factors.EBF2 belongs to the conserved Olf/EBF family (see MIM 164343) of helix-loop-helix transcription factors (Wang et al., 2002 [PubMed 12139918]).[supplied by OMIM]. PRIMARYREFSEQ_SPAN PRIMARY_IDENTIFIER PRIMARY_SPAN COMP 1-103 AU120738.1 302-404 104-650 AL701879.1 1-547 651-735 AC090103.4 79460-79544 736-796 BC069665.1 1-61 797-1794 BC074794.2 1-998 1795-2297 AK021562.1 1082-1584

Sequence

Synthetic peptide located within the following region: GDPERLAKEMLLKRAADLVEALYGTPHNNQDIILKRAADIAEALYSVPRN

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service