Skip to Content
Merck
All Photos(2)

Key Documents

PLA0248

Sigma-Aldrich

Rabbit anti-BAP1 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase), BRCA1-associated protein 1, HUCEP-13, KIAA0272, UCHL2, cerebral protein 6, cerebral protein-13, hucep-6, ubiquitin carboxy-terminal hydrolase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
R 6 513,08

R 6 513,08

List PriceR 6 646,00

Please contact Customer Service for Availability


Select a Size

Change View
100 μL
R 6 513,08

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R 6 513,08

List PriceR 6 646,00

Please contact Customer Service for Availability

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

species reactivity

human

technique(s)

immunoprecipitation (IP): 5-10 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

NP_004647.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... BAP1(8314)

Immunogen

The epitope recognized by PLA0248 maps to a region between residue 679 and 729 of human BRCA1-associated protein 1 using the numbering given in entry NP_004647.1 (GeneID 8314).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

BRCA1-associated protein 1 (BAP1) functions as a C-terminal ubiquitin hydrolase and may be involved in the transduction of signals in the BRCA1 pathway. BAP1 is also implicated to be a tumor suppressor as defects in BAP1 are found in non-small cell lung cancer cell lines.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service