Skip to Content
Merck
All Photos(1)

Key Documents

P9311

Sigma-Aldrich

Pectin, esterified potassium salt from citrus fruit

extent of labeling: 20-34% esterified

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201

biological source

citrus

form

powder

extent of labeling

20-34% esterified

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Notburga Gierlinger et al.
Planta, 227(5), 969-980 (2007-12-07)
Equisetaceae has been of research interest for decades, as it is one of the oldest living plant families, and also due to its high accumulation of silica up to 25% dry wt. Aspects of silica deposition, its association with other
Robin Eric Jacobus Spelbrink et al.
Applied biochemistry and biotechnology, 174(5), 1998-2006 (2014-08-28)
Pectin methylesterase was extracted from potato tubers and partially purified in a single chromatographic step at large industrial scale. The preparation obtained in this way matched the temperature and pH profile of the species reported earlier by Puri et al.
Túlio Ítalo S Oliveira et al.
Food chemistry, 198, 113-118 (2016-01-16)
A central composite design was used to determine effects of pH (2.0-4.5), extraction temperature (70-90 °C) and time (120-240 min) on the yield, degree of methoxylation (DM) and galacturonic acid content (GA) of pectins extracted from banana peels with citric
Renjie Li et al.
Food chemistry, 231, 96-104 (2017-04-30)
The effects of high pressure processing (HPP) on the activity of soluble acid invertase (SAI) in mango pulp, crude extract, purified SAI and purified SAI in model systems (pectin, bovine serum albumin (BSA), sugars and pH 3-7) were investigated. The
Olivier Habrylo et al.
International journal of biological macromolecules, 112, 499-508 (2018-02-01)
Pectinases represent about one fifth of the enzyme worldwide market due their wide range of biotechnological applications. Current commercial pectinases are exclusively obtained from microbial sources, but here we report a pectin methylesterase (Sl-PME) and an endo-polygalacturonase (Sl-EPG) bioprospected from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service