P7905
POPSO disodium salt
Synonym(s):
Piperazine-N,N′-bis(2-hydroxypropanesulfonic acid)
About This Item
Assay
≥99% H2SO4 basis (titration)
form
crystalline solid
useful pH range
7.2-8.5
pKa (25 °C)
7.8
solubility
water: 0.33 g/mL, clear to slightly hazy, colorless
SMILES string
OC(CS(=O)([O-])=O)CN1CCN(CC(O)CS(=O)([O-])=O)CC1.[Na+].[Na+]
InChI
1S/C10H22N2O8S2.2Na/c13-9(7-21(15,16)17)5-11-1-2-12(4-3-11)6-10(14)8-22(18,19)20;;/h9-10,13-14H,1-8H2,(H,15,16,17)(H,18,19,20);;/q;2*+1/p-2
InChI key
XMRCLLXGMGEVHS-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service