Skip to Content
Merck
All Photos(1)

Documents

P2755

Sigma-Aldrich

Propionylthiocholine chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H18ClNOS
CAS Number:
Molecular Weight:
211.75
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

−20°C

SMILES string

Cl.CCC(=O)SCC[N](C)(C)C

InChI

1S/C8H18NOS.ClH/c1-5-8(10)11-7-6-9(2,3)4;/h5-7H2,1-4H3;1H

InChI key

IBOGEBJGSGTCTA-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paola Nieri et al.
Scientific reports, 8, 45760-45760 (2017-04-04)
Organocatalysis, which is mostly explored for its new potential industrial applications, also represents a chemical event involved in endogenous processes. In the present study, we provide the first evidence that imidazole and imidazole derivatives have cholinesterase-like properties since they can
Kristina Sepčić et al.
PloS one, 14(4), e0216077-e0216077 (2019-05-01)
Cholinesterases (ChE), the enzymes whose primary function is the hydrolysis of choline esters, are widely expressed throughout the nature. Although they have already been found in plants and microorganisms, including ascomycete fungi, this study is the first report of ChE-like

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service