GE17-0044-01
Sephadex® G-50
Coarse, Cytiva, pack of 100 g
About This Item
feature
autoclavable In wet form (pH 7.0) at 120°C for 30 min
packaging
pack of 100 g
manufacturer/tradename
Cytiva
swelling
1 g swells to 9-11 mL
particle size
100-300 μm (dry)
200-610 μm (wet)
working range
2-10
InChI
1S/C18H32O16/c19-1-5(21)9(23)10(24)6(22)3-31-17-16(30)14(28)12(26)8(34-17)4-32-18-15(29)13(27)11(25)7(2-20)33-18/h1,5-18,20-30H,2-4H2
InChI key
FZWBNHMXJMCXLU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- Quickly desalts, removes contaminants and transfers to a new buffer in a single step.
- Suitable for purification of DNA from small molecules by gel filtration
- Larger bead size for large scale group separations where high flow rates and Low operating pressures are required.
- Largest bead size in the Sephadex® G-50 range.
Storage and Stability
Analysis Note
Legal Information
Storage Class Code
13 - Non Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service