Skip to Content
Merck
All Photos(2)

Key Documents

E7783

Sigma-Aldrich

p3XFLAG-CMV-13 Expression Vector

shuttle vector for transient or stable extracellular expression of C-terminal 3xFLAG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

tag

3X FLAG tagged

grade

for molecular biology

form

buffered aqueous solution

Peptide tag location

C-terminal

shipped in

dry ice

storage temp.

−20°C

General description

The p3XFLAG-CMV-13 Expression Vector is a 6.3 kb derivative of pCMV5 used to establish expression of transient or unstable C-terminal 3XFLAGfusion proteins in mammalian cells. The vector encodes three adjacent FLAG epitopes (Asp-Tyr-Lys-Xaa-Xaa-Asp) downstream of the multiple cloning region. This results in increased detection sensitivity using ANTI-FLAG?M2 antibody. Efficiency of replication and genomic integration is optimal when using an SV40 T antigen-expressing host.

p3XFLAG-CMV-7-BAP Control Plasmid is a 6.2 kb derivative of pCMV51 used for transient intracellular expression of N-terminal 3X-FLAG bacterial alkaline phosphatase fusion protein in mammalian cells. The vector encodes three adjacent FLAG epitopes (Asp- Tyr-Lys-Xaa-Xaa-Asp) upstream of the multiple cloning region. This results in increased detection sensitivity using ANTI-FLAG M2 antibody. The third FLAG epitope includes the enterokinase recognition sequence, allowing cleavage of the 3XFLAG peptide from the purified fusion protein.

Vector Maps and Sequences

Components

  • p3XFLAG-CMV-13 Expression Vector 20 μg (E4776) is supplied as 0.5 mg/ml in 10 mM Tris-HCl (pH 8.0) with 1 mM EDTA.
  • p3XFLAG-CMV-7-BAP Control Plasmid 20 μg (C7472) is supplied as 0.5 mg/ml in 10 mM Tris-HCl (pH 8.0) with 1 mM EDTA.

Principle

The promoter-regulatory region of the human cytomegalovirus drives transcription of FLAG®-fusion constructs. The preprotrypsin leader sequence precedes the FLAG® sequence.

Legal Information

This product is covered by the following patents owned by Sigma-Aldrich Co. LLC: US6,379,903, US7,094,548, JP4405125,EP1220933, CA2386471 and AU774216.
3xFLAG is a trademark of Sigma-Aldrich Co. LLC
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
p3xFLAG-CMV is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Seck et al.
The Journal of biological chemistry, 278(25), 23085-23093 (2003-04-11)
Numerous alternatively spliced transcripts are generated from the gene for the G protein-coupled calcitonin receptor, and some of the splice variants show differences in receptor-mediated signaling events. This study showed that the deltae13 splice variant of the rabbit calcitonin receptor
Seakwoo Lee et al.
The Biochemical journal, 403(1), 31-42 (2006-12-21)
Human MMP-26 (matrix metalloproteinase-26) (also known as endometase or matrilysin-2) is a putative biomarker for human carcinomas of breast, prostate and other cancers of epithelial origin. Calcium modulates protein structure and function and may act as a molecular signal or
Rong Hou et al.
The Journal of biological chemistry, 284(11), 6955-6965 (2009-01-10)
Fat1, an atypical cadherin induced robustly after arterial injury, has significant effects on mammalian vascular smooth muscle cell (VSMC) growth and migration. The related Drosophila protein Fat interacts genetically and physically with Atrophin, a protein essential for development and control
Veronica Rondahl et al.
PloS one, 8(9), e73635-e73635 (2013-09-12)
The leucine-rich repeats and immunoglobulin-like domains (LRIG) proteins constitute an integral membrane protein family that has three members: LRIG1, LRIG2, and LRIG3. LRIG1 negatively regulates growth factor signaling, but little is known regarding the functions of LRIG2 and LRIG3. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service