Rolf B1.T
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
biological source
rat nose
packaging
tube of 5 μg 03071601-DNA-5UG
pkg of vial of cells 03071601-1VL
growth mode
Adherent
karyotype
Normal rat chromosome complement
morphology
Epithelial-like
products
cadherin, tenascin, laminin, N-CAM, GFAP, p75 NTR
receptors
p75 NTR
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Adult rat olfactory nerve ensheathing cells
Cell Line Description
Rolf B1.T cells arose from a spontaneous immortalisation event - retaining characteristics of normal cells. The cells are dependent on serum growth factors for growth. They constitutively express glial fibrillary acidic protein, S100, the low affinity neurotrophin receptor p75 NTR, laminin, tenascin, and NCAM. These cells support the regrowth of neurites from adult rat retinal ganglion cells. They have potential value in research into mechanisms of glial support for axonal regeneration from adult mammalian central neurons and other aspects of differentiated glial cell biology. Epithelial-like or fusiform (inducible).
Application
Neural research including axonal regeneration.
Culture Medium
DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS)
Subculture Routine
Split sub-confluent cultures (70-80%) 1:5 - 1:20 e.g. seeding at 2-4 x 10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service